missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Tenascin C Protein
A DNA sequence encoding the Homo sapiens (Human) Tenascin C, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP8717-ec-50ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_002151.2 | |
Tenascin C Protein | |
Research Use Only | |
TNC | |
Recombinant Protein | |
E. coli | |
DSPRDLTATE VQSETALLTW RPPRASVTGY LLVYESVDGT VKEVIVGPDT TSYSLADLSP STHYTAKIQA LNGPLRSNMI QTIFTTIGLL YPFPKDCSQA MLNGDTTSGL YTIYLNGDKA EALEVFCDMT SDGGGWIVFL RRKNGRENFY QNWKAYAAGF GDRREEFWLG LDNLNKITAQ GQYELRVDLR DHGETAFAVY DKFSVGDAKT RYKLKVEGYS GTAGDSMAYH NGRSFSTFDK DTDSAITNCA LSYKGAFWYR NCHRVNLMGR YGDNNHSQGV NWFHWKGHEH SIQFAEMKLR PSNFRNLEGR RKRA | |
Greater than 90% as determined by SDS-PAGE. |
3371 | |
50 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
His | |
Tris-based buffer, 50% glycerol |