Learn More
enQuireBio™ Recombinant Human Telomerase protein component 1 Protein
A DNA sequence encoding the Homo sapiens (Human) Telomerase protein component 1, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP8918-ec-1mg
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_009041.2 | |
Telomerase protein component 1 Protein | |
1 mg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
His | |
Tris-based buffer, 50% glycerol |
7011 | |
Greater than 90% as determined by SDS-PAGE. | |
Research Use Only | |
TEP1 | |
Recombinant Protein | |
E. coli | |
LKLLCMKPGD APSEIWSSYT ENPMILSTHK EYGIFVLQPK DPGVLSFLRQ KESGEFEERL NFDINLENPS RTLISITQAK PESESSFLCA SSDGILWNLA KCSPEGEWTT GNMWQKKANT PETQTPGTDP STCRESDASM DSDASMDSEP TPHLKTRQRR KIHSGSVTAL HVLPELLVTA SKDRDVKLWE RPSMQLLGLF RCE |