missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Tektin-5 Protein
A DNA sequence encoding the Homo sapiens (Human) Tektin-5, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP8007-ec-500ug
Detalles adicionales : Peso : 0.01000kg
Specifications
NP_653275.1 | |
Tektin-5 Protein | |
Research Use Only | |
TEKT5 | |
Recombinant Protein | |
E. coli | |
MEFLGTTQTA SYCGPKKCCG LTSLPAVQAP VIQECYQPYY LPGYRYLNSW RPSLFYKIAN VQTCPDESTS TLRPPTILPT LRSALFSRYS PHDWDQSNQL QVRGAEASRL WASRLTDDSM RLLQDKDQLT HQMQEGTCRN LGQRLSDIGF WKSELSYELD RLLTENQNLE TVKRRLECAA NEVNCPLQVA LECLYHREKR IGIDLVHDNV EKNLIREVDL LKCCQEQMRK LAQRIDIQMR DNRDAQHVLE RDLEDKSSAQ CIDEKCFNLR NTSDCISFFH GMEKIDGTIS VPETWAKFSN DNIKHSQNMR ANSIQLREEA EHLFETLSDQ MWRQFTDTNL AFNARISEVT DVKNKLQTQL AKTLQEIFQA ENTIMLLERS IMAKEGPLKV AQTRLECRTR RPNMELCRDI PQLKLVNEVF TIDDTLQTLK LRLRETQDTL QLLVMTKCRL EHELAIKANT LCIDKEKCMG MRKTFPCTPR LVGHT | |
Greater than 90% as determined by SDS-PAGE. |
146279 | |
500 μg | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |