missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human TCP1 Protein
A cDNA sequence encoding the TCP1 was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP13713-10ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
TCP1 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MEGPLSVFGD RSTGETIRSQ NVMAAASIAN IVKSSLGPVG LDKMLVDDIG DVTITNDGAT ILKLLEVEHP AAKVLCELAD LQDKEVGDGT TSVVIIAAEL LKNADELVKQ KIHPTSVISG YRLACKEAVR YINENLIVNT DELGRDCLIN AAKTSMSSKI IGINGDFFAN MVVDAVLAIK YTDIRGQPRY PVNSVNILKA HGRSQMESML ISGYALNCVV GSQGMPKRIV NAKIACLDFS LQKTKMKLGV QVVITDPEKL DQIRQRESDI TKERIQKILA TGANVILTTG GIDDMCLKYF VEAGAMAVRR VLKRDLKRIA KASGATILST LANLEGEETF EAAMLGQAEE VVQERICDDE LILIKNTKAR TSASIILRGA NDFMCDEMER SLHDALCVVK RVLESKSVVP GGGAVEAALS IYLENYATSM GSREQLAIAE FARSLLVIPN TLAVNAAQDS TDLVAKLRAF HNEAQVNPER KNLKWIGLDL SNGKPRDNKQ AGVFEPTIVK VKSLKFATEA AITILRIDDL IKLHPESKDD KHGSYEDAVH SGALND | |
Greater than 80.0% as determined by SDS-PAGE. |
10 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
The TCP1 solution (0.5 mg/ml) contains 20mM Tris-HCl Buffer (pH 8.0), 1mM DTT, 0.1mM PMSF and 10% Glycerol. |