missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human TCEB1 Protein
A DNA sequence encoding the Homo sapiens (Human) TCEB1, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP6768-ec-10ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_001191786.1 | |
TCEB1 Protein | |
Research Use Only | |
TCEB1 | |
Recombinant Protein | |
E. coli | |
MDGEEKTYGG CEGPDAMYVK LISSDGHEFI VKREHALTSG TIKAMLSGPG QFAENETNEV NFREIPSHVL SKVCMYFTYK VRYTNSSTEI PEFPIAPEIA LELLMAANFL DC | |
Greater than 90% as determined by SDS-PAGE. |
6921 | |
10 μg | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |