missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Tax1-binding protein 1
A DNA sequence encoding the Homo sapiens (Human) Tax1-binding protein 3, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP8476-ec-50ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_001191627.1 | |
Tax1-binding protein 1 | |
Research Use Only | |
TAX1BP3 | |
Recombinant Protein | |
E. coli | |
MSYIPGQPVT AVVQRVEIHK LRQGENLILG FSIGGGIDQD PSQNPFSEDK TDKGIYVTRV SEGGPAEIAG LQIGDKIMQV NGWDMTMVTH DQARKRLTKR SEEVVRLLVT RQSLQKAVQQ SMLS | |
Greater than 90% as determined by SDS-PAGE. |
30851 | |
50 μg | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |