missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human TALLA-1 / TSPAN7 Protein
A DNA sequence encoding the Homo sapiens (Human) TALLA-1 / TSPAN7, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP6846-ec-500ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_004606.2 | |
TALLA-1 / TSPAN7 Protein | |
Research Use Only | |
TSPAN7 | |
Recombinant Protein | |
E. coli | |
RHEIKDTFLR TYTDAMQTYN GNDERSRAVD HVQRSLSCCG VQNYTNWSTS PYFLEHGIPP SCCMNETDCN PQDLHNLTVA ATKVNQKGCY DLVTSFMETN M | |
Greater than 80.90% as determined by SDS-PAGE. |
7102 | |
500 μg | |
Not determined. | |
Extracellular Domain | |
Human | |
His | |
Tris-based buffer, 50% glycerol |