missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human T-complex protein 1 subunit beta Protein
A DNA sequence encoding the Homo sapiens (Human) T-complex protein 1 subunit beta, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP5779-ec-500ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_001185771.1 | |
T-complex protein 1 subunit beta Protein | |
Research Use Only | |
CCT2 | |
Recombinant Protein | |
E. coli | |
ASLSLAPVNI FKAGADEERA ETARLTSFIG AIAIGDLVKS TLGPKGMDKI LLSSGRDASL MVTNDGATIL KNIGVDNPAA KVLVDMSRVQ DDEVGDGTTS VTVLAAELLR EAESLIAKKI HPQTIIAGWR EATKAAREAL LSSAVDHGSD EVKFRQDLMN IAGTTLSSKL LTHHKDHFTK LAVEAVLRLK GSGNLEAIHI IKKLGGSLAD SYLDEGFLLD KKIGVNQPKR IENAKILIAN TGMDTDKIKI FGSRVRVDST AKVAEIEHAE KEKMKEKVER ILKHGINCFI NRQLIYNYPE QLFGAAGVMA IEHADFAGVE RLALVTGGEI ASTFDHPELV KLGSCKLIEE VMIGEDKLIH FSGVALGEAC TIVLRGATQQ ILDEAERSLH DALCVLAQTV KDSRTVYGGG CSEMLMAHAV TQLANRTPGK EAVAMESYAK ALRMLPTIIA DNAGYDSADL VAQLRAAHSE GNTTAGLDMR EGTIGDMAIL GITESFQVKR QVLLSAAEAA EVILRVDNII KAAPRKRVPD HHPC | |
Greater than 90% as determined by SDS-PAGE. |
10576 | |
500 μg | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |