missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human SULT2B1 Protein
A DNA sequence encoding the Homo sapiens (Human) SULT2B1, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP6749-ec-10ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_004596.2 | |
SULT2B1 Protein | |
Research Use Only | |
SULT2B1 | |
Recombinant Protein | |
E. coli | |
MDGPAEPQIP GLWDTYEDDI SEISQKLPGE YFRYKGVPFP VGLYSLESIS LAENTQDVRD DDIFIITYPK SGTTWMIEII CLILKEGDPS WIRSVPIWER APWCETIVGA FSLPDQYSPR LMSSHLPIQI FTKAFFSSKA KVIYMGRNPR DVVVSLYHYS KIAGQLKDPG TPDQFLRDFL KGEVQFGSWF DHIKGWLRMK GKDNFLFITY EELQQDLQGS VERICGFLGR PLGKEALGSV VAHSTFSAMK ANTMSNYTLL PPSLLDHRRG AFLRKGVCGD WKNHFTVAQS EAFDRAYRKQ MRGMPTFPWD EDPEEDGSPD PEPSPEPEPK PSLEPNTSLE REPRPNSSPS PSPGQASETP HPRPS | |
Greater than 80% as determined by SDS-PAGE. |
6820 | |
10 μg | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |