missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Substance-P receptor Protein
A DNA sequence encoding the Homo sapiens (Human) Substance-P receptor, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP8669-ec-200ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_001049.1 | |
Substance-P receptor Protein | |
Research Use Only | |
NK1R | |
Recombinant Protein | |
E. coli | |
MDNVLPVDSD LSPNISTNTS EPNQFVQPA | |
Greater than 90% as determined by SDS-PAGE. |
6869 | |
200 μg | |
Not determined. | |
Extracellular Domain | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |