missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human SUB1 Protein
A DNA sequence encoding the Homo sapiens (Human) SUB1, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP6745-ec-10ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_006704.3 | |
SUB1 Protein | |
Research Use Only | |
SUB1 | |
Recombinant Protein | |
E. coli | |
PKSKELVSSS SSGSDSDSEV DKKLKRKKQV APEKPVKKQK TGETSRALSS SKQSSSSRDD NMFQIGKMRY VSVRDFKGKV LIDIREYWMD PEGEMKPGRK GISLNPEQWS QLKEQISDID DAVRKL | |
Greater than 90% as determined by SDS-PAGE. |
10923 | |
10 μg | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |