missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human STRAP Protein
A DNA sequence encoding the Homo sapiens (Human) STRAP, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP8281-ec-200ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_009109.3 | |
STRAP Protein | |
Research Use Only | |
STRAP | |
Recombinant Protein | |
E. coli | |
MAMRQTPLTC SGHTRPVVDL AFSGITPYGY FLISACKDGK PMLRQGDTGD WIGTFLGHKG AVWGATLNKD ATKAATAAAD FTAKVWDAVS GDELMTLAHK HIVKTVDFTQ DSNYLLTGGQ DKLLRIYDLN KPEAEPKEIS GHTSGIKKAL WCSEDKQILS ADDKTVRLWD HATMTEVKSL NFNMSVSSME YIPEGEILVI TYGRSIAFHS AVSLDPIKSF EAPATINSAS LHPEKEFLVA GGEDFKLYKY DYNSGEELES YKGHFGPIHC VRFSPDGELY ASGSEDGTLR LWQTVVGKTY GLWKCVLPEE DSGELAKPKI GFPETTEEEL EEIASENSDC IFPSAPDVKA | |
Greater than 80% as determined by SDS-PAGE. |
11171 | |
200 μg | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |