missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human STK3 Protein
A cDNA sequence encoding the STK3 was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP13619-10ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
STK3 Protein | |
Research Use Only | |
Recombinant Protein | |
Baculovirus | |
ADPMEQPPAP KSKLKKLSED SLTKQPEEVF DVLEKLGEGS YGSVFKAIHK ESGQVVAIKQ VPVESDLQEI IKEISIMQQC DSPYVVKYYG SYFKNTDLWI VMEYCGAGSV SDIIRLRNKT LIEDEIATIL KSTLKGLEYL HFMRKIHRDI KAGNILLNTE GHAKLADFGV AGQLTDTMAK RNTVIGTPFW MAPEVIQEIG YNCVADIWSL GITSIEMAEG KPPYADIHPM RAIFMIPTNP PPTFRKPELW SDDFTDFVKK CLVKNPEQRA TATQLLQHPF IKNAKPVSIL RDLITEAMEI KAKRHEEQQR ELEEEEENSD EDELDHHHHH H | |
Greater than 90% as determined by SDS-PAGE. |
10 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
STK3 protein solution (0.25 mg/ml) containing Phosphate Buffered Saline (pH 7.4) and 10% glycerol. |