missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human STARD5 Protein
A cDNA sequence encoding the STARD5 was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP13610-1mg
Detalles adicionales : Peso : 0.01000kg
Especificaciones
STARD5 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMDPALAA QMSEAVAEKM LQYRRDTAGW KICREGNGVS VSWRPSVEFP GNLYRGEGIV YGTLEEVWDC VKPAVGGLRV KWDENVTGFE IIQSITDTLC VSRTSTPSAA MKLISPRDFV DLVLVKRYED GTISSNATHV EHPLCPPKPG FVRGFNHPCG CFCEPLPGEP TKTNLVTFFH TDLSGYLPQN VVDSFFPRSM TRFYANLQKA VKQFHE | |
Greater than 95.0% as determined by SDS-PAGE. |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
STARD5 protein solution (1 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0), 0.1M NaCl, 1mM DTT and 20% glycerol. |