missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human SSX1 Protein
A cDNA sequence encoding the SSX1 was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP13600-1mg
Detalles adicionales : Peso : 0.01000kg
Especificaciones
SSX1 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMNGDDTF AKRPRDDAKA SEKRSKAFDD IATYFSKKEW KKMKYSEKIS YVYMKRNYKA MTKLGFKVTL PPFMCNKQAT DFQGNDFDND HNRRIQVEHP QMTFGRLHRI IPKIMPKKPA EDENDSKGVS EASGPQNDGK QLHPPGKANI SEKINKRSGP KRGKHAWTHR LRERKQLVIY EEISDPEEDD E | |
Greater than 85.0% as determined by SDS-PAGE. |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
SSX1 protein solution (1 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0), 0.4M Urea and 10% glycerol. |