missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Squalene synthase Protein
A DNA sequence encoding the Homo sapiens (Human) Squalene synthase, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP6031-ec-10ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_001274671.1 | |
Squalene synthase Protein | |
Research Use Only | |
FDFT1 | |
Recombinant Protein | |
E. coli | |
EFVKCLGHPE EFYNLVRFRI GGKRKVMPKM DQDSLSSSLK TCYKYLNQTS RSFAAVIQAL DGEMRNAVCI FYLVLRALDT LEDDMTISVE KKVPLLHNFH SFLYQPDWRF MESKEKDRQV LEDFPTISLE FRNLAEKYQT VIADICRRMG IGMAEFLDKH VTSEQEWDKY CHYVAGLVGI GLSRLFSASE FEDPLVGEDT ERANSMGLFL QKTNIIRDYL EDQQGGREFW PQEVWSRYVK KLGDFAKPEN IDLAVQCLNE LITNALHHIP DVITYLSRLR NQSVFNFCAI PQVMAIATLA ACYNNQQVFK GAVKIRKGQA VTLMMDATNM PAVKAIIYQY MEEIYHRIPD SDPSSSKTRQ IISTIRTQNL PNCQLISRSH YSPIYLSFVM LLAALSWQYL TTLSQVTEDY VQTGEH | |
Greater than 90% as determined by SDS-PAGE. |
2222 | |
10 μg | |
Not determined. | |
Full Length | |
Human | |
His | |
Tris-based buffer, 50% glycerol |