missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Sperm-egg fusion protein Juno Protein
A DNA sequence encoding the Homo sapiens (Human) Sperm-egg fusion protein Juno, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP6052-ec-50ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_001186135.1 | |
Sperm-egg fusion protein Juno Protein | |
Research Use Only | |
IZUMO1R | |
Recombinant Protein | |
E. coli | |
GDELLNICMN AKHHKRVPSP EDKLYEECIP WKDNACCTLT TSWEAHLDVS PLYNFSLFHC GLLMPGCRKH FIQAICFYEC SPNLGPWIQP VGSLGWEVAP SGQGERVVNV PLCQEDCEEW WEDCRMSYTC KSNWRGGWDW SQGKNRCPKG AQCLPFSHYF PTPADLCEKT WSNSFKASPE RRNSGRCLQK WFEPAQGNPN VAVARLFASS APSWELSYTI MVCSLFLPFL S | |
Greater than 90% as determined by SDS-PAGE. |
390243 | |
50 μg | |
Not determined. | |
Full Length | |
Human | |
His | |
Tris-based buffer, 50% glycerol |