missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human SOS1 Protein
A cDNA sequence encoding the SOS1 was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP13560-5ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
SOS1 Protein | |
Research Use Only | |
Recombinant Protein | |
Baculovirus | |
ADPEEQMRLP SADVYRFAEP DSEENIIFEE NMQPKAGIPI IKAGTVIKLI ERLTYHMYAD PNFVRTFLTT YRSFCKPQEL LSLIIERFEI PEPEPTEADR IAIENGDQPL SAELKRFRKE YIQPVQLRVL NVCRHWVEHH FYDFERDAYL LQRMEEFIGT VRGKAMKKWV ESITKIIQRK KIARDNGPGH NITFQSSPPT VEWHISRPGH IETFDLLTLH PIEIARQLTL LESDLYRAVQ PSELVGSVWT KEDKEINSPN LLKMIRHTTN LTLWFEKCIV ETENLEERVA VVSRIIEILQ VFQELNNFNG VLEVVSAMNS SPVYRLDHTF EQIPSRQKKI LEEAHELSED HYKKYLAKLR SINPPCVPFF GIYLTNILKT EEGNPEVLKR HGKELINFSK RRKVAEITGE IQQYQNQPYC LRVESDIKRF FENLNPMGNS MEKEFTDYLF NKSLEIEPRN PKPLPRFPKK YSYPLKSPGV RPSNPRPGTH HHHHH | |
Greater than 95.0% as determined by SDS-PAGE. |
5 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
SOS1 protein solution (0.25 mg/ml) contains 30% glycerol, 0.1M NaCl, 1mM DTT and 0.2mM MgCl2. |