missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human SNRPD3 Protein
A cDNA sequence encoding the SNRPD3 was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP13548-HIS-INSECT-10ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
SNRPD3 Protein | |
Research Use Only | |
Recombinant Protein | |
Baculovirus | |
MGSSHHHHHH SSGLVPRGSH MSIGVPIKVL HEAEGHIVTC ETNTGEVYRG KLIEAEDNMN CQMSNITVTY RDGRVAQLEQ VYIRGSKIRF LILPDMLKNA PMLKSMKNKN QGSGAGRGKA AILKAQVAAR GRGRGMGRGN IFQKRR | |
Greater than 85% as determined by SDS-PAGE. |
10 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
The SNRPD3 solution (0.25 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 0.5M NaCl, 2mM DTT, 0.1mM PMSF and 40% glycerol. |