missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human SMAC/DIABLO Protein
A cDNA sequence encoding the SMAC/DIABLO was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP13525-20ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
SMAC/DIABLO Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MASMTGGQQM GRGSMAVPIA QKSEPHSLSS EALMRRAVSL VTDSTSTFLS QTTYALIEAI TEYTKAVYTL TSLYRQYTSL LGKMNSEEED EVWQVIIGAR AEMTSKHQEY LKLETTWMTA VGLSEMAAEA AYQTGADQAS ITARNHIQLV KLQVEEVHQL SRKAETKLAE AQIEELRQKT QEEGEERAES EQEAYLRED | |
Greater than 95.0% as determined by:(a) Analysis by RP-HPLC. (b) Analysis by SDS-PAGE. |
20 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
Untagged | |
The Smac/Diablo solution contains 20mM Tris pH 7.5. |