missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human SLC34A2 Protein
A DNA sequence encoding the Homo sapiens (Human) SLC34A2, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP9403-ye-10ug
Additional Details : Peso : 0.01000kg
Specifications
NP_001171469.1 | |
SLC34A2 Protein | |
Research Use Only | |
SLC34A2 | |
Recombinant Protein | |
Yeast | |
LLQSRCPRVL PKKLQNWNFL PLWMRSLKPW DAVVSKFTGC FQMRCCCCCR VCCRACCLLC DCPKCCRCSK CCEDLEEAQE GQDVPVKAPE TFDNITISRE AQGEVPASDS KTECTA | |
Greater than 90% as determined by SDS-PAGE. |
10568 | |
10 μg | |
Not determined. | |
Cytoplasmic Domain | |
Human | |
His | |
Tris-based buffer, 50% glycerol |