missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human SLAMF6 Protein
A cDNA sequence encoding the SLAMF6 was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP13519-HIS-20ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
SLAMF6 Protein | |
20 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
SLAMF6 protein solution (1 mg/ml) contains 20mM Tris-HCl buffer, (pH 8.0), 0.4 Urea and 10% glycerol. |
Greater than 85.0% as determined by SDS-PAGE. | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSQSSLTPL MVNGILGESV TLPLEFPAGE KVNFITWLFN ETSLAFIVPH ETKSPEIHVT NPKQGKRLNF TQSYSLQLSN LKMEDTGSYR AQISTKTSAK LSSYTLRILR QLRNIQVTNH SQLFQNMTCE LHLTCSVEDA DDNVSFRWEA LGNTLSSQPN LTVSWDPRIS SEQDYTCIAE NAVSNLSFSV SAQKLCEDVK IQYTDTKM |