missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human SH3BGRL3 Protein
A cDNA sequence encoding the SH3BGRL3 was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP13489-20ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
SH3BGRL3 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMSGLRVY STSVTGSREI KSQQSEVTRI LDGKRIQYQL VDISQDNALR DEMRALAGNP KATPPQIVNG DQYCGDYELF VEAVEQNTLQ EFLKLA | |
Greater than 95% as determined by SDS-PAGE. |
20 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
The SH3BGRL3 solution (1 mg/1ml) contains 20mM Tris-HCl buffer (pH 8.0), 0.15M NaCl, 1mM DTT and 10% glycerol. |