missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Sesquipedalian-2 Protein
A DNA sequence encoding the Homo sapiens (Human) Sesquipedalian-2, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP7760-ec-1mg
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_001002034.2 | |
Sesquipedalian-2 Protein | |
1 mg | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
150368 | |
Greater than 90% as determined by SDS-PAGE. | |
Research Use Only | |
FAM109B | |
Recombinant Protein | |
E. coli | |
MKLNERSVAH YALSDSPADH MGFLRTWGGP GTPPTPSGTG RRCWFVLKGN LLFSFESREG RAPLSLVVLE GCTVELAEAP VPEEFAFAIC FDAPGVRPHL LAAEGPAAQE AWVKVLSRAS FGYMRLVVRE LESQLQDARQ SLALQRRSSW KSVASRCKPQ APNHRAAGLE NGHCLSKDSS PVGLVEEAGS RSAGWGLAEW ELQGPASLLL GKGQSPVSPE TSCFSTLHDW YGQEIVELRQ CWQKRAQGSH SKCEEQDRP |