missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Serine / arginine-rich splicing factor 1 Protein
A DNA sequence encoding the Homo sapiens (Human) Serine/arginine-rich splicing factor 1, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP6679-ec-10ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_001071634.1 | |
Serine / arginine-rich splicing factor 1 Protein | |
Research Use Only | |
SRSF1 | |
Recombinant Protein | |
E. coli | |
SGGGVIRGPA GNNDCRIYVG NLPPDIRTKD IEDVFYKYGA IRDIDLKNRR GGPPFAFVEF EDPRDAEDAV YGRDGYDYDG YRLRVEFPRS GRGTGRGGGG GGGGGAPRGR YGPPSRRSEN RVVVSGLPPS GSWQDLKDHM REAGDVCYAD VYRDGTGVVE FVRKEDMTYA VRKLDNTKFR SHEGETAYIR VKVDGPRSPS YGRSRSRSRS RSRSRSRSNS RSRSYSPRRS RGSPRYSPRH SRSRSRT | |
Greater than 90% as determined by SDS-PAGE. |
6426 | |
10 μg | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |