missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Septin-7 Protein
A DNA sequence encoding the Homo sapiens (Human) Septin-7, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP7576-ec-50ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_001779.3 | |
Septin-7 Protein | |
Research Use Only | |
42985 | |
Recombinant Protein | |
E. coli | |
SVSARSAAAE ERSVNSSTMV AQQKNLEGYV GFANLPNQVY RKSVKRGFEF TLMVVGESGL GKSTLINSLF LTDLYSPEYP GPSHRIKKTV QVEQSKVLIK EGGVQLLLTI VDTPGFGDAV DNSNCWQPVI DYIDSKFEDY LNAESRVNRR QMPDNRVQCC LYFIAPSGHG LKPLDIEFMK RLHEKVNIIP LIAKADTLTP EECQQFKKQI MKEIQEHKIK IYEFPETDDE EENKLVKKIK DRLPLAVVGS NTIIEVNGKR VRGRQYPWGV AEVENGEHCD FTILRNMLIR THMQDLKDVT NNVHYENYRS RKLAAVTYNG VDNNKNKGQL TKSPLAQMEE ERREHVAKMK KMEMEMEQVF EMKVKEKVQK LKDSEAELQR RHEQMKKNLE AQHKELEEKR RQFEDEKANW EAQQRILEQQ NSSRTLEKNK KKGKIF | |
Greater than 90% as determined by SDS-PAGE. |
989 | |
50 μg | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |