missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Selenide, water dikinase 1 Protein
A DNA sequence encoding the Homo sapiens (Human) Selenide, water dikinase 1, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP6663-ec-200ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_001182531.1 | |
Selenide, water dikinase 1 Protein | |
Research Use Only | |
SEPHS1 | |
Recombinant Protein | |
E. coli | |
STRESFNPES YELDKSFRLT RFTELKGTGC KVPQDVLQKL LESLQENHFQ EDEQFLGAVM PRLGIGMDTC VIPLRHGGLS LVQTTDYIYP IVDDPYMMGR IACANVLSDL YAMGVTECDN MLMLLGVSNK MTDRERDKVM PLIIQGFKDA AEEAGTSVTG GQTVLNPWIV LGGVATTVCQ PNEFIMPDNA VPGDVLVLTK PLGTQVAVAV HQWLDIPEKW NKIKLVVTQE DVELAYQEAM MNMARLNRTA AGLMHTFNAH AATDITGFGI LGHAQNLAKQ QRNEVSFVIH NLPVLAKMAA VSKACGNMFG LMHGTCPETS GGLLICLPRE QAARFCAEIK SPKYGEGHQA WIIGIVEKGN RTARIIDKPR IIEVAPQVAT QNVNPTPGAT S | |
Greater than 80% as determined by SDS-PAGE. |
22929 | |
200 μg | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |