missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Scrapie-responsive protein 1
A DNA sequence encoding the Homo sapiens (Human) Scrapie-responsive protein 1, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP7501-ye-10ug
Detalles adicionales : Peso : 0.01000kg
Specifications
NP_001316526.1 | |
Scrapie-responsive protein 1 | |
Research Use Only | |
SCRG1 | |
Recombinant Protein | |
Yeast | |
MPANRLSCYR KILKDHNCHN LPEGVADLTQ IDVNVQDHFW DGKGCEMICY CNFSELLCCP KDVFFGPKIS FVIPCNNQ | |
Greater than 90% as determined by SDS-PAGE. |
11341 | |
10 μg | |
Not determined. | |
Full Length | |
Human | |
His | |
Tris-based buffer, 50% glycerol |