missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human SCAMP3 Protein
A DNA sequence encoding the Homo sapiens (Human) SCAMP3, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP8499-ec-200ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_005689.2 | |
SCAMP3 Protein | |
Research Use Only | |
SCAMP3 | |
Recombinant Protein | |
E. coli | |
AQSRDGGNPF AEPSELDNPF QDPAVIQHRP SRQYATLDVY NPFETREPPP AYEPPAPAPL PPPSAPSLQP SRKLSPTEPK NYGSYSTQAS AAAATAELLK KQEELNRKAE ELDRRERELQ HAALGGTATR QNNWPPLPSF CPVQPCFFQD ISMEIPQEFQ KTVSTMYYL | |
Greater than 90% as determined by SDS-PAGE. |
10067 | |
200 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |