missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human S100A5 Protein
A cDNA sequence encoding the S100A5 was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP13396-20ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
S100A5 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH METPLEKALT TMVTTFHKYS GREGSKLTLS RKELKELIKK ELCLGEMKES SIDDLMKSLD KNSDQEIDFK EYSVFLTMLC MAYNDFFLED NK | |
Greater than 90.0% as determined by SDS-PAGE. |
20 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
The S100A5 solution (1 mg/1ml) contains 20mM Tris-HCl buffer (pH 8.0), 1mM DTT, 30% glycerol, 0.1M NaCl and 0.1mM PMSF. |