missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human RPS7 Protein
A cDNA sequence encoding the RPS7 was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP13360-1mg
Detalles adicionales : Peso : 0.01000kg
Especificaciones
RPS7 Protein | |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
RPS7 protein solution (0.5 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0), 100mM NaCl, 1mM DTT and 30% glycerol. |
Greater than 90% as determined by SDS-PAGE. | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSHMFSSSA KIVKPNGEKP DEFESGISQA LLELEMNSDL KAQLRELNIT AAKEIEVGGG RKAIIIFVPV PQLKSFQKIQ VRLVRELEKK FSGKHVVFIA QRRILPKPTR KSRTKNKQKR PRSRTLTAVH DAILEDLVFP SEIVGKRIRV KLDGSRLIKV HLDKAQQNNV EHKVETFSGV YKKLTGKDVN FEFPEFQL |