missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human RPP25L Protein
A DNA sequence encoding the Homo sapiens (Human) RPP25L, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP7851-ec-10ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_680544.1 | |
RPP25L Protein | |
Research Use Only | |
RPP25L | |
Recombinant Protein | |
E. coli | |
MEHYRKAGSV ELPAPSPMPQ LPPDTLEMRV RDGSKIRNLL GLALGRLEGG SARHVVFSGS GRAAGKAVSC AEIVKRRVPG LHQLTKLRFL QTEDSWVPAS PDTGLDPLTV RRHVPAVWVL LSRDPLDPNE CGYQPPGAPP GLGSMPSSSC GPRSRRRARD TRS | |
Greater than 80% as determined by SDS-PAGE. |
138716 | |
10 μg | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |