missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human RPL7A Protein
A cDNA sequence encoding the RPL7A was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP13348-10ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
RPL7A Protein | |
10 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
RPL7A protein solution (1 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0) and 10% glycerol. |
Greater than 85.0% as determined by SDS-PAGE. | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMPKGKKA KGKKVAPAPA VVKKQEAKKV VNPLFEKRPK NFGIGQDIQP KRDLTRFVKW PRYIRLQRQR AILYKRLKVP PAINQFTQAL DRQTATQLLK LAHKYRPETK QEKKQRLLAR AEKKAAGKGD VPTKRPPVLR AGVNTVTTLV ENKKAQLVVI AHDVDPIELV VFLPALCRKM GVPYCIIKGK ARLGRLVHRK TCTTVAFTQV NSEDKGALAK LVEAIRTNYN DRYDEIRRHW GGNVLGPKSV ARIAKLEKAK AKELATKLG |