missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human RNF4 Protein
A cDNA sequence encoding the RNF4 was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP13331-20ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
RNF4 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMSTRKRR GGAINSRQAQ KRTREATSTP EISLEAEPIE LVETAGDEIV DLTCESLEPV VVDLTHNDSV VIVDERRRPR RNARRLPQDH ADSCVVSSDD EELSRDRDVY VTTHTPRNAR DEGATGLRPS GTVSCPICMD GYSEIVQNGR LIVSTECGHV FCSQCLRDSL KNANTCPTCR KKINHKRYHP IYI | |
Greater than 90.0% as determined by SDS-PAGE. |
20 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
RNF4 protein solution (0.5 mg/ml) containing 20mM Tris-HCl buffer (pH 8.5), 0.2M NaCl, 30% glycerol and 1mM DTT. |