missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human RIPK2 Protein
A DNA sequence encoding the Homo sapiens (Human) RIPK2, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP6609-ec-200ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_003812.1 | |
RIPK2 Protein | |
Research Use Only | |
RIPK2 | |
Recombinant Protein | |
E. coli | |
MNGEAICSAL PTIPYHKLAD LRYLSRGASG TVSSARHADW RVQVAVKHLH IHTPLLDSER KDVLREAEIL HKARFSYILP ILGICNEPEF LGIVTEYMPN GSLNELLHRK TEYPDVAWPL RFRILHEIAL GVNYLHNMTP PLLHHDLKTQ NILLDNEFHV KIADFGLSKW RMMSLSQSRS SKSAPEGGTI IYMPPENYEP GQKSRASIKH DIYSYAVITW EVLSRKQPFE DVTNPLQIMY SVSQGHRPVI NEESLPYDIP HRARMISLIE SGWAQNPDER PSFLKCLIEL EPVLRTFEEI TFLEAVIQLK KTKLQSVSSA IHLCDKKKME LSLNIPVNHG PQEESCGSSQ LHENSGSPET SRSLPAPQDN DFLSRKAQDC YFMKLHHCPG NHSWDSTISG SQRAAFCDHK TTPCSSAIIN PLSTAGNSER LQPGIAQQWI QSKREDIVNQ MTEACLNQSL DALLSRDLIM KEDYELVSTK PTRTSKVRQL LDTTDIQGEE FAKVIVQKLK DNKQMGLQPY PEILVVSRSP SLNLLQNKSM | |
Greater than 90% as determined by SDS-PAGE. |
8767 | |
200 μg | |
Not determined. | |
Full Length | |
Human | |
His | |
Tris-based buffer, 50% glycerol |