missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human RING-box protein 1
A DNA sequence encoding the Homo sapiens (Human) RING-box protein 2, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP6613-ye-10ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_001188299.1 | |
RING-box protein 1 | |
Research Use Only | |
RNF7 | |
Recombinant Protein | |
Yeast | |
ADVEDGEETC ALASHSGSSG SKSGGDKMFS LKKWNAVAMW SWDVECDTCA ICRVQVMDAC LRCQAENKQE DCVVVWGECN HSFHNCCMSL WVKQNNRCPL CQQDWVVQRI GK | |
Greater than 90% as determined by SDS-PAGE. |
9616 | |
10 μg | |
Not determined. | |
Full Length | |
Human | |
His | |
Tris-based buffer, 50% glycerol |