missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human RGS21 Protein
A cDNA sequence encoding the RGS21 was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP13311-20ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
RGS21 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSHMPVKCC FYRSPTAETM TWSENMDTLL ANQAGLDAFR IFLKSEFSEE NVEFWLACED FKKTKNADKI ASKAKMIYSE FIEADAPKEI NIDFGTRDLI SKNIAEPTLK CFDEAQKLIY CLMAKDSFPR FLKSEIYKKL VNSQQVPNHK KWLPFL | |
Greater than 90% as determined by SDS-PAGE. |
20 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
RGS21 protein solution (1 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0), 1mM DTT, 30% glycerol and 0.1M NaCl. |