missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Reticulocalbin-1 Protein
A DNA sequence encoding the Homo sapiens (Human) Reticulocalbin-1, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP9756-ye-10ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_002892.1 | |
Reticulocalbin-1 Protein | |
Research Use Only | |
RCN1 | |
Recombinant Protein | |
Yeast | |
PTVRKERVVR PDSELGERPP EDNQSFQYDH EAFLGKEDSK TFDQLTPDES KERLGKIVDR IDNDGDGFVT TEELKTWIKR VQKRYIFDNV AKVWKDYDRD KDDKISWEEY KQATYGYYLG NPAEFHDSSD HHTFKKMLPR DERRFKAADL NGDLTATREE FTAFLHPEEF EHMKEIVVLE TLEDIDKNGD GFVDQDEYIA DMFSHEENGP EPDWVLSERE QFNEFRDLNK DGKLDKDEIR HWILPQDYDH AQAEARHLVY ESDKNKDEKL TKEEILENWN MFVGSQATNY GEDLTKNHDE L | |
Greater than 90% as determined by SDS-PAGE. |
5954 | |
10 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
His | |
Tris-based buffer, 50% glycerol |