Learn More
enQuireBio™ Recombinant Human REG1A Protein
A cDNA sequence encoding the REG1A was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP13292-10ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
REG1A Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MKHHHHHHAS HMQEAQTELP QARISCPEGT NAYRSYCYYF NEDRETWVDA DLYCQNMNSG NLVSVLTQAE GAFVASLIKE SGTDDFNVWI GLHDPKKNRR WHWSSGSLVS YKSWGIGAPS SVNPGYCVSL TSSTGFQKWK DVPCEDKFSF VCKFKN | |
Greater than 95% as determined by SDS-PAGE. |
10 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
Filtered (0.4microm) and lyophilized from 0.5 mg/ml in 5mM Tris, 25mM NaCl, pH 7.5. |