missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human RBP7 Protein
A cDNA sequence encoding the RBP7 was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP13285-20ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
RBP7 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MPADLSGTWT LLSSDNFEGY MLALGIDFAT RKIAKLLKPQ KVIEQNGDSF TIHTNSSLRN YFVKFKVGEE FDEDNRGLDN RKCKSLVIWD NDRLTCIQKG EKKNRGWTHW IEGDKLHLEM FCEGQVCKQT FQRA | |
Greater than 90.0% as determined by SDS-PAGE. |
20 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
The RBP7 solution (0.5 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 0.2M NaCl, 2mM DTT and 20% glycerol. |