missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Ras-related protein Rap-2b Protein
A DNA sequence encoding the Homo sapiens (Human) Ras-related protein Rap-2b, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP6584-ec-50ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_002877.2 | |
Ras-related protein Rap-2b Protein | |
Research Use Only | |
RAP2B | |
Recombinant Protein | |
E. coli | |
MREYKVVVLG SGGVGKSALT VQFVTGSFIE KYDPTIEDFY RKEIEVDSSP SVLEILDTAG TEQFASMRDL YIKNGQGFIL VYSLVNQQSF QDIKPMRDQI IRVKRYERVP MILVGNKVDL EGEREVSYGE GKALAEEWSC PFMETSAKNK ASVDELFAEI VRQMNYAAQP NGDEGCCSAC | |
Greater than 80% as determined by SDS-PAGE. |
5912 | |
50 μg | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |