missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Ras-related protein Rab-5A Protein
A DNA sequence encoding the Homo sapiens (Human) Ras-related protein Rab-5A, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP6571-ec-500ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_001278977.1 | |
Ras-related protein Rab-5A Protein | |
Research Use Only | |
RAB5A | |
Recombinant Protein | |
E. coli | |
MASRGATRPN GPNTGNKICQ FKLVLLGESA VGKSSLVLRF VKGQFHEFQE STIGAAFLTQ TVCLDDTTVK FEIWDTAGQE RYHSLAPMYY RGAQAAIVVY DITNEESFAR AKNWVKELQR QASPNIVIAL SGNKADLANK RAVDFQEAQS YADDNSLLFM ETSAKTSMNV NEIFMAIAKK LPKNEPQNPG ANSARGRGVD LTEPTQPTRN QCCSN | |
Greater than 90% as determined by SDS-PAGE. |
5868 | |
500 μg | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |