missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human RAB32 Protein
A cDNA sequence encoding the RAB32 was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP13237-1mg
Detalles adicionales : Peso : 0.01000kg
Especificaciones
RAB32 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSHMAGGGA GDPGLGAAAA PAPETREHLF KVLVIGELGV GKTSIIKRYV HQLFSQHYRA TIGVDFALKV LNWDSRTLVR LQLWDIAGQE RFGNMTRVYY KEAVGAFVVF DISRSSTFEA VLKWKSDLDS KVHLPNGSPI PAVLLANKCD QNKDSSQSPS QVDQFCKEHG FAGWFETSAK DNINIEEAAR FLVEKILVNH QSFPNEENDV DKIKLDQETL RAENKSQCC | |
Greater than 90% as determined by SDS-PAGE. |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
The RAB32 solution (0.5 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 5mM DTT, 0.2M NaCl, 0.2M EDTA and 50% glycerol. |