missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human RAB27B Protein
A DNA sequence encoding the Homo sapiens (Human) RAB27B, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP6568-ec-10ug
Detalles adicionales : Peso : 0.01000kg
Spécification
NP_004154.2 | |
RAB27B Protein | |
Research Use Only | |
RAB27B | |
Recombinant Protein | |
E. coli | |
TDGDYDYLIK LLALGDSGVG KTTFLYRYTD NKFNPKFITT VGIDFREKRV VYNAQGPNGS SGKAFKVHLQ LWDTAGQERF RSLTTAFFRD AMGFLLMFDL TSQQSFLNVR NWMSQLQANA YCENPDIVLI GNKADLPDQR EVNERQAREL ADKYGIPYFE TSAATGQNVE KAVETLLDLI MKRMEQCVEK TQIPDTVNGG NSGNLDGEKP PEKKCIC | |
Greater than 90% as determined by SDS-PAGE. |
5874 | |
10 μg | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |