missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human PSMB8 Protein
A cDNA sequence encoding the PSMB8 was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP13190-1mg
Detalles adicionales : Peso : 0.01000kg
Especificaciones
PSMB8 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSHMTTTLA FKFQHGVIAA VDSRASAGSY ISALRVNKVI EINPYLLGTM SGCAADCQYW ERLLAKECRL YYLRNGERIS VSAASKLLSN MMCQYRGMGL SMGSMICGWD KKGPGLYYVD EHGTRLSGNM FSTGSGNTYA YGVMDSGYRP NLSPEEAYDL GRRAIAYATH RDSYSGGVVN MYHMKEDGWV KVESTDVSDL LHQYREANQ | |
Greater than 85.0% as determined by SDS-PAGE. |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
PSMB8 protein solution (0.5 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 0.4M Urea and 10% glycerol. |