missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human PSMB5 Protein
A cDNA sequence encoding the PSMB5 was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP13187-1mg
Detalles adicionales : Peso : 0.01000kg
Especificaciones
PSMB5 Protein | |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
The PSMB5 solution (0.25 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 20% glycerol, 5mM DTT and 0.2M NaCl. |
Greater than 90.0% as determined by SDS-PAGE. | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MRGSHHHHHH GMASMTGGQQ MGRDLYDDDD KDRWGSMTTT LAFKFRHGVI VAADSRATAG AYIASQTVKK VIEINPYLLG TMAGGAADCS FWERLLARQC RIYELRNKER ISVAAASKLL ANMVYQYKGM GLSMGTMICG WDKRGPGLYY VDSEGNRISG ATFSVGSGSV YAYGVMDRGY SYDLEVEQAY DLARRAIYQA TYRDAYSGGA VNLYHVREDG WIRVSSDNVA DLHEKYSGST P |