missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Protein Mpv17 Protein
A DNA sequence encoding the Homo sapiens (Human) Protein Mpv17, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP6377-ec-200ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_002428.1 | |
Protein Mpv17 Protein | |
Research Use Only | |
MPV17 | |
Recombinant Protein | |
E. coli | |
MALWRAYQRA LAAHPWKVQV LTAGSLMGLG DIISQQLVER RGLQEHQRGR TLTMVSLGCG FVGPVVGGWY KVLDRFIPGT TKVDALKKML LDQGGFAPCF LGCFLPLVGA LNGLSAQDNW AKLQRDYPDA LITNYYLWPA VQLANFYLVP LHYRLAVVQC VAVIWNSYLS WKAHRL | |
Greater than 90% as determined by SDS-PAGE. |
4358 | |
200 μg | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |