missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Protein FAM110B Protein
A DNA sequence encoding the Homo sapiens (Human) Protein FAM110B, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP6018-ec-100ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_671722.1 | |
Protein FAM110B Protein | |
Research Use Only | |
FAM110B | |
Recombinant Protein | |
E. coli | |
MPTETLQTGS MVKPVSPAGT FTSAVPLRIL NKGPDYFRRQ AEPNPKRLSA VERLEADKAK YVKSQEVINA KQEPVKPAVL AKPPVCPAAK RALGSPTLKV FGNHAKTESG VQRENLKLEI LKNIINSSEG SSSGSGHKHS SRNWPPHRSE ATDLHRHSFA ESLKVYPTQG RRSPQEGGSH VGRRLLEQSA ESFLHVSHSS SDIRKVTSVK PLKAIPCSSS APPLPPKPKI AAIASMKSPE ADPVEPACGV SRRPSLQRSK SDLSDRYFRV DADVERFFNY CGLDPEELEN LGMENFARAN SDIISLNFRS ASMISSDCEQ SQDSNSDLRN DDSANDRVPY GISAIERNAR IIKWLYSIKQ ARESQKVSHV | |
Greater than 90% as determined by SDS-PAGE. |
90362 | |
100 μg | |
Not determined. | |
Full Length | |
Human | |
His | |
Tris-based buffer, 50% glycerol |