missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Protein delta homolog 2 Protein
A DNA sequence encoding the Homo sapiens (Human) Protein delta homolog 2, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP7755-ye-50ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_001273584.1 | |
Protein delta homolog 2 Protein | |
Research Use Only | |
DLK2 | |
Recombinant Protein | |
Yeast | |
DDCSSHCDLA HGCCAPDGSC RCDPGWEGLH CERCVRMPGC QHGTCHQPWQ CICHSGWAGK FCDKDEHICT TQSPCQNGGQ CMYDGGGEYH CVCLPGFHGR DCERKAGPCE QAGSPCRNGG QCQDDQGFAL NFTCRCLVGF VGARCEVNVD DCLMRPCANG ATCLDGINRF SCLCPEGFAG RFCTINLDDC ASRPCQRGAR CRDRVHDFDC LCPSGYGGKT CELVLPVPDP PTTVDTPLGP TSAVVVPATG PAPHSAGAGL LRISVKEVVR RQEAGLGEPS | |
Greater than 90% as determined by SDS-PAGE. |
65989 | |
50 μg | |
Not determined. | |
Extracellular Domain | |
Human | |
His | |
Tris-based buffer, 50% glycerol |