missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Protein delta homolog 1 Protein
A DNA sequence encoding the Homo sapiens (Human) Protein delta homolog 1, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP5937-ec-200ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_001304101.1 | |
Protein delta homolog 1 Protein | |
Research Use Only | |
DLK1 | |
Recombinant Protein | |
E. coli | |
AECFPACNPQ NGFCEDDNVC RCQPGWQGPL CDQCVTSPGC LHGLCGEPGQ CICTDGWDGE LCDRDVRACS SAPCANNRTC VSLDDGLYEC SCAPGYSGKD CQKKDGPCVI NGSPCQHGGT CVDDEGRASH ASCLCPPGFS GNFCEIVANS CTPNPCENDG VCTDIGGDFR CRCPAGFIDK TCSRPVTNCA SSPCQNGGTC LQHTQVSYEC LCKPEFTGLT CVKKRALSPQ QVTRLPSGYG LAYRLTPGVH ELPVQQPEHR ILKVSMKELN KKTPLLTEGQ | |
Greater than 90% as determined by SDS-PAGE. |
8788 | |
200 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
His | |
Tris-based buffer, 50% glycerol |